missing translation for 'onlineSavingsMsg'
Learn More

5-Azaindole, 98%, Acros Organics™

Brand:  Acros Organics 456660010

Additional Details : CAS : 271-34-1 Weight : 0.00100kg


Quantity 1g

 View more versions of this product

Product Code. 15505206

  • 67.40€ / 1g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

271-34-1
118.139
5-azaindole, 1h-pyrrolo 3,2-c pyridine, 1,5-diazaindene, 3,7-dideazapurine, 1h-pyrrolo 3,2-c pyridine, 5-azindole, 5h-pyrrolo 3,2-c pyridine, pyrrolo 3,2-c pyridine, 5-aza indole, 5-aza-indole
1H-pyrrolo[3,2-c]pyridine
C7H6N2
SRSKXJVMVSSSHB-UHFFFAOYSA-N
9220
C1=CNC2=C1C=NC=C2

Specifications

5-Azaindole
93.0°C to 94.0°C
271-34-1
5-azaindole, 1h-pyrrolo 3,2-c pyridine, 1,5-diazaindene, 3,7-dideazapurine, 1h-pyrrolo 3,2-c pyridine, 5-azindole, 5h-pyrrolo 3,2-c pyridine, pyrrolo 3,2-c pyridine, 5-aza indole, 5-aza-indole
C1=CNC2=C1C=NC=C2
118.139
118.14
Authentic
1g
C7H6N2
SRSKXJVMVSSSHB-UHFFFAOYSA-N
1H-pyrrolo[3,2-c]pyridine
9220
≥97.5% (HPLC)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Azaindole, 98%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.