missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindole hydrochloride, 95%, Thermo Scientific™

Catalog Number p-3770788
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
65795-92-8
C8H8N2·HCl
168.63
MFCD00077647
CZRIFWIECZRIKN-UHFFFAOYSA-N
5-aminoindole hydrochloride, 1h-indol-5-amine hydrochloride, 5-aminoindole monohydrochloride, 5-aminoindole hcl, 5-aminoindol-hydrochloride, 5-indolamine hydrochloride, 1h-indol-5-amine,hydrochloride
16211959
1H-indol-5-amine;hydrochloride
C1=CC2=C(C=CN2)C=C1N.Cl
This item is not returnable. View return policy
65795-92-8
C8H8N2·HCl
168.63
MFCD00077647
CZRIFWIECZRIKN-UHFFFAOYSA-N
5-aminoindole hydrochloride, 1h-indol-5-amine hydrochloride, 5-aminoindole monohydrochloride, 5-aminoindole hcl, 5-aminoindol-hydrochloride, 5-indolamine hydrochloride, 1h-indol-5-amine,hydrochloride
16211959
1H-indol-5-amine;hydrochloride
C1=CC2=C(C=CN2)C=C1N.Cl
CAS Min % 94.0
CAS Max % 100.0
Melting Point 253.0°C
Color Brown to Tan
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 168.63
Percent Purity 95%
Physical Form Crystals
Chemical Name or Material 5-Aminoindole hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 5-Aminoindole hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'