Learn More
5,6,7,8-Tetrahydroquinoline, 98%, Thermo Scientific Chemicals
Catalog Number p-7077190
Click to view available options
Quantity:
5 g
25 g
100 g
10500-57-9
C9H11N
133.19
YQDGQEKUTLYWJU-UHFFFAOYSA-N
2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
66335
5,6,7,8-tetrahydroquinoline
C1CCC2=C(C1)C=CC=N2
10500-57-9
C9H11N
133.19
YQDGQEKUTLYWJU-UHFFFAOYSA-N
2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
66335
5,6,7,8-tetrahydroquinoline
C1CCC2=C(C1)C=CC=N2
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.0300g/mL
Boiling Point 218.0°C to 222.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Refractive Index 1.544
Quantity 5 g
Beilstein 20,II,176
Specific Gravity 1.03
Formula Weight 133.19
Percent Purity 98%
Chemical Name or Material 5,6,7,8-Tetrahydroquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 5,6,7,8-Tetrahydroquinoline, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.