missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydroquinoline, 98%, Acros Organics™

Brand:  Acros Organics 188870250

Additional Details : CAS : 10500-57-9 Weight : 0.02500kg


Quantity 25g

 View more versions of this product

Product Code. 15505876

  • 78.30€ / 25g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

10500-57-9
133.194
2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
5,6,7,8-tetrahydroquinoline
C9H11N
YQDGQEKUTLYWJU-UHFFFAOYSA-N
66335
C1CCC2=C(C1)C=CC=N2

Specifications

5,6,7,8-Tetrahydroquinoline
86°C
1.544
218.0°C to 222.0°C
10500-57-9
2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
C1CCC2=C(C1)C=CC=N2
133.194
133.19
1.0300g/mL
Authentic
1.03
25g
C9H11N
YQDGQEKUTLYWJU-UHFFFAOYSA-N
5,6,7,8-tetrahydroquinoline
66335
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5,6,7,8-Tetrahydroquinoline, 98%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.