missing translation for 'onlineSavingsMsg'
Learn More

5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one, 97%, Thermo Scientific Chemicals

Catalog Number p-7039563
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
851116-03-5
C11H8N2OS2
248.318
MFCD06671220
SYSIILLIYRVOMM-UHFFFAOYSA-N
5-5-methylthiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-5-methylthiophen-2-yl-3h,4h-thieno 2,3-d pyrimidin-4-one, 5-5-methyl-2-thienyl thieno 2,3-d pyrimidin-4-one
4963645
5-(5-methylthiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one
CC1=CC=C(S1)C2=CSC3=C2C(=O)NC=N3
This item is not returnable. View return policy
851116-03-5
C11H8N2OS2
248.318
MFCD06671220
SYSIILLIYRVOMM-UHFFFAOYSA-N
5-5-methylthiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-5-methylthiophen-2-yl-3h,4h-thieno 2,3-d pyrimidin-4-one, 5-5-methyl-2-thienyl thieno 2,3-d pyrimidin-4-one
4963645
5-(5-methylthiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one
CC1=CC=C(S1)C2=CSC3=C2C(=O)NC=N3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 248.31
Percent Purity 97%
Chemical Name or Material 5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'