Learn More
KGW Isotherm™ 4000mL Dewar Carrying Flask with Lid
Brand: KGW Isotherm™ 1224E
Additional Details : Weight : 0.80000kg
Specifications
Insulated lid, held by two spring clips, handle | |
Cooling of small samples | |
305mm | |
Borosilicate Glass |
138 mm | |
310mm | |
160mm | |
4 L |