missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Tri-n-butylstannyl)thiazole, 95%, Thermo Scientific Chemicals
53.60 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Quantity | 250 mg |
UN Number | UN2788 |
Solubility Information | Not miscible in water. |
IUPAC Name | tributyl(1,3-thiazol-4-yl)stannane |
Formula Weight | 374.17 |
Percent Purity | 95% |
Chemical Name or Material | 4-(Tri-n-butylstannyl)thiazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'