missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexylamine, 97%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number p-7077566
27.86 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 mL
25 mL
This item is not returnable. View return policy
5400-88-4
C10H21N
155.29
MFCD00013763,MFCD20690546
BGNLXETYTAAURD-UHFFFAOYSA-N
4-tert-butylcyclohexylamine, 4-tert-butyl cyclohexanamine, 4-t-butylcyclohexylamine, 4-tert-butylcyclohexanamine, 4-amino-1-tert-butylcyclohexane, 4-tert.-butylcyclohexylamine, 4-tert-butylcyclohexyl amine, cyclohexanamine, 4-1,1-dimethylethyl, cis-1-amino-4-tert-butylcyclohexane, 4-tert-butylcyclohexylamine,c&t
79396
4-tert-butylcyclohexan-1-amine
CC(C)(C)C1CCC(N)CC1
This item is not returnable. View return policy
5400-88-4
C10H21N
155.29
MFCD00013763,MFCD20690546
BGNLXETYTAAURD-UHFFFAOYSA-N
4-tert-butylcyclohexylamine, 4-tert-butyl cyclohexanamine, 4-t-butylcyclohexylamine, 4-tert-butylcyclohexanamine, 4-amino-1-tert-butylcyclohexane, 4-tert.-butylcyclohexylamine, 4-tert-butylcyclohexyl amine, cyclohexanamine, 4-1,1-dimethylethyl, cis-1-amino-4-tert-butylcyclohexane, 4-tert-butylcyclohexylamine,c&t
79396
4-tert-butylcyclohexan-1-amine
CC(C)(C)C1CCC(N)CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8700g/mL
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (total of isomers) (GC)
Quantity 5 mL
Specific Gravity 0.87
Solubility Information Solubility in water: immiscible.
Formula Weight 155.29
Percent Purity 97%
Chemical Name or Material 4-tert-Butylcyclohexylamine
compliance-icons
Product Identifier
  • 4-tert-Butylcyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'