missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butyl)phenyl isocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Density | 0.986g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)3CC6H4NCO |
Quantity | 1 g |
Formula Weight | 175.23 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Tert-butyl)phenyl isocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'