missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Piperidinobenzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Synonym | 4-piperidin-1-yl-benzoic acid, 4-piperidin-1-yl benzoic acid, 4-piperidinobenzoic acid, 4-piperidinobenzenecarboxylic acid, benzoic acid, 4-1-piperidinyl, 1-4-carboxyphenyl piperidine, 4-1-piperidinyl benzoic acid, benzoic acid,4-1-piperidinyl, 4-piperidylbenzoic acid, cambridge id 5267560 |
Formula Weight | 205.26 |
Percent Purity | 97% |
Chemical Name or Material | 4-Piperidinobenzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'