missing translation for 'onlineSavingsMsg'
Learn More

[4-(Phenoxymethyl)phenyl]methanol, 97%, Thermo Scientific™

Catalog Number p-3586562
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
262862-97-5
C14H14O2
214.264
MFCD09879941
MTQHORUPINPVTP-UHFFFAOYSA-N
4-phenoxymethyl phenyl methanol, 4-phenoxymethylphenylmethanol, 4-phenoxymethylbenzyl alcohol, 4-phenoxymethylphenyl-methanol, 4-phenoxymethyl phenyl methan-1-ol
11333367
[4-(phenoxymethyl)phenyl]methanol
C1=CC=C(C=C1)OCC2=CC=C(C=C2)CO
This item is not returnable. View return policy
262862-97-5
C14H14O2
214.264
MFCD09879941
MTQHORUPINPVTP-UHFFFAOYSA-N
4-phenoxymethyl phenyl methanol, 4-phenoxymethylphenylmethanol, 4-phenoxymethylbenzyl alcohol, 4-phenoxymethylphenyl-methanol, 4-phenoxymethyl phenyl methan-1-ol
11333367
[4-(phenoxymethyl)phenyl]methanol
C1=CC=C(C=C1)OCC2=CC=C(C=C2)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 214.26
Percent Purity 97%
Chemical Name or Material [4-(Phenoxymethyl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'