missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl phosphate, 99%, Thermo Scientific Chemicals

Catalog Number p-7043189
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
3368-04-5
MFCD00016969
This item is not returnable. View return policy
3368-04-5
MFCD00016969

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 214°C to 216°C
Color White
Molecular Formula C10H9O6P
Quantity 250 mg
Beilstein 1687229
Synonym 4-methylumbelliferyl phosphate, 4-methyl-2-oxo-2h-chromen-7-yl dihydrogen phosphate, 4-mup, 2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy, phosphoric acid 4-methylumbelliferyl ester, 4-methylumbelliferylphosphate, free acid, 4-methyl-2-oxochromen-7-yl oxyphosphonic acid, 5,5-dinitrobarbutyric acid, 4-methylumbelliferone phosphate, 4-methylumbelliferyl phosphate, free acid
Solubility Information Soluble in water
InChI Key BCHIXGBGRHLSBE-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)(O)O
IUPAC Name (4-methyl-2-oxochromen-7-yl) dihydrogen phosphate
Molecular Weight (g/mol) 256.15
PubChem CID 65118
Formula Weight 256.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'