missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzaldehyde, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-115754
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
100-10-7
C9H11NO
149.19
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
100-10-7
C9H11NO
149.19
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 73°C to 75°C
Color White to Off-white
Density 5.15
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 25 g
Beilstein 14, 31
Fieser 01,273; 02,146
Merck Index 15, 3252
Solubility Information Solubility in water: 0.3g/L (20°C) Other solubilities: Soluble in acetic acid, Soluble in alcohol, ether and chloroform
Formula Weight 149.19
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'