Learn More
4-(Cyanomethoxy)phenylboronic acid, 97%, ACROS Organics™
Brand: Acros Organics 445400050
Additional Details : CAS : 947533-23-5 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
947533-23-5 | |
176.97 | |
4-cyanomethoxyphenylboronic acid, 4-cyanomethoxy phenyl boronic acid, 4-cyanomethoxy phenylboronic acid, acmc-209rtj | |
[4-(cyanomethoxy)phenyl]boronic acid |
C8H8BNO3 | |
GKJPNYHKHSLHCC-UHFFFAOYSA-N | |
21918429 | |
B(C1=CC=C(C=C1)OCC#N)(O)O |
Specifications
4-(Cyanomethoxy)phenylboronic acid | |
947533-23-5 | |
C8H8BNO3 | |
GKJPNYHKHSLHCC-UHFFFAOYSA-N | |
[4-(cyanomethoxy)phenyl]boronic acid | |
21918429 | |
97% |
5g | |
96% min. (HPLC) | |
4-cyanomethoxyphenylboronic acid, 4-cyanomethoxy phenyl boronic acid, 4-cyanomethoxy phenylboronic acid, acmc-209rtj | |
B(C1=CC=C(C=C1)OCC#N)(O)O | |
176.97 | |
176.97 |
Safety and Handling
- 4-(Cyanomethoxy)phenylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.