missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-90820
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
1878-66-6
C8H7ClO2
170.59
MFCD00004344
CDPKJZJVTHSESZ-UHFFFAOYSA-N
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
15880
CHEBI:30749
2-(4-chlorophenyl)acetic acid
OC(=O)CC1=CC=C(Cl)C=C1
This item is not returnable. View return policy
1878-66-6
C8H7ClO2
170.59
MFCD00004344
CDPKJZJVTHSESZ-UHFFFAOYSA-N
4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
15880
CHEBI:30749
2-(4-chlorophenyl)acetic acid
OC(=O)CC1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 103°C to 107°C
CAS Min % 97.5
Color White to Cream
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula ClC6H4CH2CO2H
Beilstein 09,448
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,solubility 5% in chcl3: clear solution
Formula Weight 170.6
Percent Purity 98%
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Chemical Name or Material 4-Chlorophenylacetic acid, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'