missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-2-methyl-1,3-thiazole hydrochloride , Tech., Thermo Scientific™

Catalog Number p-4509946
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
77470-53-2
C5H7Cl2NS
184.08
MFCD00067725
YGKDISJLDVGNOR-UHFFFAOYSA-N
4-chloromethyl-2-methylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-methylthiazole hcl, 4-chloromethyl-2-methyl thiazole hydrochloride, 2-methyl-4-chloromethyl thiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole, chloride, 2-methyl-4-chloromethylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hcl
2734203
4-(chloromethyl)-2-methyl-1,3-thiazole;hydrochloride
Cl.CC1=NC(CCl)=CS1
This item is not returnable. View return policy
77470-53-2
C5H7Cl2NS
184.08
MFCD00067725
YGKDISJLDVGNOR-UHFFFAOYSA-N
4-chloromethyl-2-methylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-methylthiazole hcl, 4-chloromethyl-2-methyl thiazole hydrochloride, 2-methyl-4-chloromethyl thiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole, chloride, 2-methyl-4-chloromethylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hcl
2734203
4-(chloromethyl)-2-methyl-1,3-thiazole;hydrochloride
Cl.CC1=NC(CCl)=CS1
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 184.09
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Chloromethyl)-2-methyl-1, 3-thiazole hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'