missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Chlorochalcone, 98+%
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Specifications
Specifications
| Melting Point | 114°C to 116°C |
| Quantity | 10 g |
| Beilstein | 1105953 |
| Formula Weight | 242.71 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Chlorochalcone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction