Learn More
4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific Chemicals
Catalog Number p-3770930
29.93 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
938-71-6
C7H5Cl2NO2
206.02
MFCD00007216
QBIQAXWMONJLNW-UHFFFAOYSA-N
4-chloro-2-nitrobenzyl chloride, 4-chloro-1-chloromethyl-2-nitrobenzene, 4-chloro-2-nitrobenzylchloride, acmc-20amlj, benzene, 4-chloro-1-chloromethyl-2-nitro, 2-nitro-4-chlorobenzyl chloride, omphlgrocarzou-uhfffaoysa, .alpha.,4-dichloro-2-nitrotoluene
598399
[O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1
938-71-6
C7H5Cl2NO2
206.02
MFCD00007216
QBIQAXWMONJLNW-UHFFFAOYSA-N
4-chloro-2-nitrobenzyl chloride, 4-chloro-1-chloromethyl-2-nitrobenzene, 4-chloro-2-nitrobenzylchloride, acmc-20amlj, benzene, 4-chloro-1-chloromethyl-2-nitro, 2-nitro-4-chlorobenzyl chloride, omphlgrocarzou-uhfffaoysa, .alpha.,4-dichloro-2-nitrotoluene
598399
[O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 38°C to 44°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H3(NO2)CH2Cl
Quantity 1 g
IUPAC Name 4-chloro-1-(chloromethyl)-2-nitrobenzene
Formula Weight 206.03
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-nitrobenzyl chloride
compliance-icons
Product Identifier
  • 4-Chloro-2-nitrobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.