missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoronitrobenzene, 98%, Thermo Scientific™

Catalog Number p-7077591
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
700-37-8
C6H3ClFNO2
175.54
MFCD00042211
PTCPUGKKWNMITF-UHFFFAOYSA-N
4-chloro-2-fluoronitrobenzene, 2-fluoro-4-chloronitrobenzene, 1-chloro-3-fluoro-4-nitrobenzene, benzene, 4-chloro-2-fluoro-1-nitro, 4-chloro-2-fluoro-1-nitro-benzene, 4-chlor-2-fluor-1-nitrobenzol, pubchem1051, acmc-1bl8e, 4-chloro2-fluoronitrobenzene, ksc380m1b
69691
4-chloro-2-fluoro-1-nitrobenzene
[O-][N+](=O)C1=CC=C(Cl)C=C1F
This item is not returnable. View return policy
700-37-8
C6H3ClFNO2
175.54
MFCD00042211
PTCPUGKKWNMITF-UHFFFAOYSA-N
4-chloro-2-fluoronitrobenzene, 2-fluoro-4-chloronitrobenzene, 1-chloro-3-fluoro-4-nitrobenzene, benzene, 4-chloro-2-fluoro-1-nitro, 4-chloro-2-fluoro-1-nitro-benzene, 4-chlor-2-fluor-1-nitrobenzol, pubchem1051, acmc-1bl8e, 4-chloro2-fluoronitrobenzene, ksc380m1b
69691
4-chloro-2-fluoro-1-nitrobenzene
[O-][N+](=O)C1=CC=C(Cl)C=C1F
Melting Point 48.0°C to 50.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 175.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoronitrobenzene
compliance-icons
Product Identifier
  • 4-Chloro-2-fluoronitrobenzene, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'