missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrophenol, 98%, Thermo Scientific™

Catalog Number p-2690590
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7693-52-9
C6H4BrNO3
218.01
MFCD00082540
CUTFAPGINUFNQM-UHFFFAOYSA-N
2-nitro-4-bromophenol, phenol, 4-bromo-2-nitro, 4-bromo-2-nitro-phenol, 4-bromo-1-hydroxy-2-nitrobenzene, phenol, 2-nitro-5-bromo, pubchem4099, 4-brom-2-nitro-phenol, acmc-209p6y, 4-06-00-01363 beilstein handbook reference, ksc494e2j
24364
4-bromo-2-nitrophenol
OC1=CC=C(Br)C=C1[N+]([O-])=O
This item is not returnable. View return policy
7693-52-9
C6H4BrNO3
218.01
MFCD00082540
CUTFAPGINUFNQM-UHFFFAOYSA-N
2-nitro-4-bromophenol, phenol, 4-bromo-2-nitro, 4-bromo-2-nitro-phenol, 4-bromo-1-hydroxy-2-nitrobenzene, phenol, 2-nitro-5-bromo, pubchem4099, 4-brom-2-nitro-phenol, acmc-209p6y, 4-06-00-01363 beilstein handbook reference, ksc494e2j
24364
4-bromo-2-nitrophenol
OC1=CC=C(Br)C=C1[N+]([O-])=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 87.0°C to 94.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(NO2)OH
Quantity 5 g
Beilstein 06, IV, 1363
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,toluene
Formula Weight 218.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromo-2-nitrophenol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'