missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxyphenol, 98%, Thermo Scientific Chemicals

Catalog Number p-4323338
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7368-78-7
C7H7BrO2
203.04
WHSIIJQOEGXWSN-UHFFFAOYSA-N
4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
262234
4-bromo-2-methoxyphenol
COC1=C(C=CC(=C1)Br)O
This item is not returnable. View return policy
7368-78-7
C7H7BrO2
203.04
WHSIIJQOEGXWSN-UHFFFAOYSA-N
4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
262234
4-bromo-2-methoxyphenol
COC1=C(C=CC(=C1)Br)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 35°C to 39°C
Color Brown to Colorless
Boiling Point 90°C to 92°C (1.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 203.04
Percent Purity 98%
Physical Form Low Melting Solid or Crystalline Solid
Chemical Name or Material 4-Bromo-2-methoxyphenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-methoxyphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'