Learn More
4-Bromo-2-fluorobenzyl cyanide, 98+%, Thermo Scientific™
Catalog Number p-3212014
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
114897-91-5
C8H5BrFN
214.04
MFCD07368653
QLASQEZPJFNZQC-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl cyanide, 2-4-bromo-2-fluorophenyl acetonitrile, 4-bromo-2-fluorophenylacetonitrile, 2-fluoro-4-bromophenylacetonitrile, benzeneacetonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorophenyl acetonitrile, 2-4-bromo-2-fluorophenyl ethanenitrile, 4-bromo-2-fluorobenzylcyanide, pubchem10413, acmc-2099lv
7172308
2-(4-bromo-2-fluorophenyl)acetonitrile
FC1=CC(Br)=CC=C1CC#N
114897-91-5
C8H5BrFN
214.04
MFCD07368653
QLASQEZPJFNZQC-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl cyanide, 2-4-bromo-2-fluorophenyl acetonitrile, 4-bromo-2-fluorophenylacetonitrile, 2-fluoro-4-bromophenylacetonitrile, benzeneacetonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorophenyl acetonitrile, 2-4-bromo-2-fluorophenyl ethanenitrile, 4-bromo-2-fluorobenzylcyanide, pubchem10413, acmc-2099lv
7172308
2-(4-bromo-2-fluorophenyl)acetonitrile
FC1=CC(Br)=CC=C1CC#N
CAS Min % 98.0
CAS Max % 100.0
Melting Point 52.8°C to 53.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CNCH2C6H3(F)Br
Quantity 5 g
Formula Weight 214.04
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzyl cyanide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-fluorobenzyl cyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.