missing translation for 'onlineSavingsMsg'
Learn More

4-[b-(5-Methylpyrazinyl-2-carboxamido)ethyl]benzene Sulfonamide, TRC

Product Code. 30466331
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466331 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466331 Supplier Toronto Research Chemicals Supplier No. TRCM3259462.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 33288-71-0
Molecular Formula C14 H16 N4 O3 S
Quantity 2.5 g
Synonym 2-Pyrazinecarboxamide, N-[2-[4-(aminosulfonyl)phenyl]ethyl]-5-methyl-, Pyrazinecarboxamide, 5-methyl-N-(p-sulfamoylphenethyl)- (8CI), Pyrazinecarboxamide, N-[2-[4-(aminosulfonyl)phenyl]ethyl]-5-methyl- (9CI), N-[2-[4-(Aminosulfonyl)phenyl]ethyl]-5-methyl-2-pyrazinecarboxamide, 4-[2-(5-Methylpyrazine-2-carboxamido)ethyl]benzenesulfonamide, 4-[2-(5-Methylpyrazinyl-2-carboxamido)ethyl]benzenesulfonamide, 5-Methyl-N-[2-(4-sulfamoylphenyl)ethyl]pyrazine-2-carboxamide, 5-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide, 4-[β-(5-Methylpyrazinyl-2-carboxamido)ethyl]benzene Sulfonamide, N-{2-[(4-Aminosulfonyl)phenyl]ethyl}-5-methylpyrazinecarboxamide
InChI Formula InChI=1S/C14H16N4O3S/c1-10-8-18-13(9-17-10)14(19)16-7-6-11-2-4-12(5-3-11)22(15,20)21/h2-5,8-9H,6-7H2,1H3,(H,16,19)(H2,15,20,21)
SMILES Cc1cnc(cn1)C(=O)NCCc2ccc(cc2)S(=O)(=O)N
IUPAC Name 5-methyl-N-[2-(4-sulfamoylphenyl)ethyl]pyrazine-2-carboxamide
Molecular Weight (g/mol) 320.37
Formula Weight 320.0943
Chemical Name or Material 5-Methyl-N-[2-(4-sulfamoylphenyl)ethyl]pyrazine-2-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.