missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidoacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-3593310
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2719-21-3
C10H11NO2
177.2
MFCD00014965
WECHHDJTILFYQT-UHFFFAOYSA-N
4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
75937
N-(4-acetylphenyl)acetamide
CC(=O)C1=CC=C(C=C1)NC(=O)C
This item is not returnable. View return policy
2719-21-3
C10H11NO2
177.2
MFCD00014965
WECHHDJTILFYQT-UHFFFAOYSA-N
4'-acetamidoacetophenone, n-4-acetylphenyl acetamide, 4-acetamidoacetophenone, n-p-acetylphenyl acetamide, acetamide, n-4-acetylphenyl, p-acetylaminoacetophenone, 4-acetylacetanilide, 4'-acetylacetanilide, p-acetamidoacetophenone, 4-acetamido-acetophenone
75937
N-(4-acetylphenyl)acetamide
CC(=O)C1=CC=C(C=C1)NC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 166°C to 170°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3COC6H4NHCOCH3
Quantity 5 g
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in benzene
Formula Weight 177.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetamidoacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'