missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Bromopyrimidin-2-yl)morpholine, 97%, Thermo Scientific Chemicals

Catalog Number p-4228467
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
84539-22-0
C8H10BrN3O
244.09
MFCD00483251
CEXBOCXKAGPRHD-UHFFFAOYSA-N
4-5-bromopyrimidin-2-yl morpholine, 5-bromo-2-morpholin-1-yl-pyrimidine, 4-5-bromo-pyrimidin-2-yl-morpholine, morpholine, 4-5-bromo-2-pyrimidinyl, 5-bromo-2-morpholinopyrimidine, 5-bromo-2-4-morpholino pyrimidine, 5-bromo-2-morpholin-4-yl pyrimidine, acmc-209pvb, 2-morpholino-5-bromopyrimidine, 5-bromo-2-morpholin-1-ylpyrimidine
5144362
4-(5-bromopyrimidin-2-yl)morpholine
C1COCCN1C2=NC=C(C=N2)Br
This item is not returnable. View return policy
84539-22-0
C8H10BrN3O
244.09
MFCD00483251
CEXBOCXKAGPRHD-UHFFFAOYSA-N
4-5-bromopyrimidin-2-yl morpholine, 5-bromo-2-morpholin-1-yl-pyrimidine, 4-5-bromo-pyrimidin-2-yl-morpholine, morpholine, 4-5-bromo-2-pyrimidinyl, 5-bromo-2-morpholinopyrimidine, 5-bromo-2-4-morpholino pyrimidine, 5-bromo-2-morpholin-4-yl pyrimidine, acmc-209pvb, 2-morpholino-5-bromopyrimidine, 5-bromo-2-morpholin-1-ylpyrimidine
5144362
4-(5-bromopyrimidin-2-yl)morpholine
C1COCCN1C2=NC=C(C=N2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 91°C to 93°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 244.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(5-Bromopyrimidin-2-yl)morpholine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(5-Bromopyrimidin-2-yl)morpholine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'