missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Isocyanatophenoxy)tetrahydropyran, 97%, Thermo Scientific™

Catalog Number p-3541066
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892501-94-9
C12H13NO3
219.24
MFCD09702366
ZGNHUEWFJQLPDN-UHFFFAOYSA-N
4-4-isocyanatophenoxy oxane, 4-4-isocyanatophenoxy tetrahydropyran, 4-4-isocyanatophenoxy tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-4-isocyanatophenoxy, 4-tetrahydro-2h-pyran-4-yl oxy phenyl isocyanate, 1-isocyanato-4-tetrahydro-2h-pyran-4-yl oxy benzene, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenisocyanate
24229521
4-(4-isocyanatophenoxy)oxane
C1COCCC1OC2=CC=C(C=C2)N=C=O
This item is not returnable. View return policy
892501-94-9
C12H13NO3
219.24
MFCD09702366
ZGNHUEWFJQLPDN-UHFFFAOYSA-N
4-4-isocyanatophenoxy oxane, 4-4-isocyanatophenoxy tetrahydropyran, 4-4-isocyanatophenoxy tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-4-isocyanatophenoxy, 4-tetrahydro-2h-pyran-4-yl oxy phenyl isocyanate, 1-isocyanato-4-tetrahydro-2h-pyran-4-yl oxy benzene, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenisocyanate
24229521
4-(4-isocyanatophenoxy)oxane
C1COCCC1OC2=CC=C(C=C2)N=C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material 4-(4-Isocyanatophenoxy)tetrahydropyran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'