missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluoropiperidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number 15505186
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
144230-52-4
C5H9F2N·HCl
157.59
OABUKBBBSMNNPM-UHFFFAOYSA-N
4,4-difluoropiperidine hydrochloride, 4,4-difluoropiperidine hcl, piperidine, 4,4-difluoro-, hydrochloride, 4,4-difluoropiperidine, chloride, 4,4-difluoropiperidin-1-ium chloride, 4,4-difluoropiperidinehydrochloride, pubchem6797, acmc-1bwk7, 4,4-difluoropiperidinehcl, 4,4-difluoropiperidin hcl
2758351
4,4-difluoropiperidine;hydrochloride
C1CNCCC1(F)F.Cl
This item is not returnable. View return policy
144230-52-4
C5H9F2N·HCl
157.59
OABUKBBBSMNNPM-UHFFFAOYSA-N
4,4-difluoropiperidine hydrochloride, 4,4-difluoropiperidine hcl, piperidine, 4,4-difluoro-, hydrochloride, 4,4-difluoropiperidine, chloride, 4,4-difluoropiperidin-1-ium chloride, 4,4-difluoropiperidinehydrochloride, pubchem6797, acmc-1bwk7, 4,4-difluoropiperidinehcl, 4,4-difluoropiperidin hcl
2758351
4,4-difluoropiperidine;hydrochloride
C1CNCCC1(F)F.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 173.0°C to 177.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 104% (Argentometry)
Quantity 5 g
Formula Weight 157.59
Percent Purity 97%
Chemical Name or Material 4,4-Difluoropiperidine hydrochloride
compliance-icons
Product Identifier
  • 4,4-Difluoropiperidine hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'