missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2700647
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
95233-37-7
C13H15ClO2
238.711
MFCD06797626
NXXDIEYTMQYWJU-UHFFFAOYSA-N
4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
2735792
4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O
This item is not returnable. View return policy
95233-37-7
C13H15ClO2
238.711
MFCD06797626
NXXDIEYTMQYWJU-UHFFFAOYSA-N
4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
2735792
4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Linear Formula ClC6H4C6H10CO2H
Formula Weight 238.71
Percent Purity 97%
Quantity 1 g
Packaging Amber Glass Bottle
Chemical Name or Material (E)-4-(4-Chlorophenyl)cyclohexanecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'