missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(2-Methylphenoxy)phenylsulfonylaminomethyl]benzoic acid, 96%, Thermo Scientific™

Catalog Number p-7032516
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
885268-78-0
C21H19NO5S
397.445
MFCD06409346
RAJRZONWHZRZEN-UHFFFAOYSA-N
4-4-o-tolyloxy phenyl sulfonamido methyl benzoic acid, 4-4-o-tolyloxy phenylsulfonamido methyl benzoic acid, 4-4-2-methylphenoxy benzenesulfonamido methyl benzoic acid
44669129
4-[[[4-(2-methylphenoxy)phenyl]sulfonylamino]methyl]benzoic acid
CC1=CC=CC=C1OC2=CC=C(C=C2)S(=O)(=O)NCC3=CC=C(C=C3)C(=O)O
This item is not returnable. View return policy
885268-78-0
C21H19NO5S
397.445
MFCD06409346
RAJRZONWHZRZEN-UHFFFAOYSA-N
4-4-o-tolyloxy phenyl sulfonamido methyl benzoic acid, 4-4-o-tolyloxy phenylsulfonamido methyl benzoic acid, 4-4-2-methylphenoxy benzenesulfonamido methyl benzoic acid
44669129
4-[[[4-(2-methylphenoxy)phenyl]sulfonylamino]methyl]benzoic acid
CC1=CC=CC=C1OC2=CC=C(C=C2)S(=O)(=O)NCC3=CC=C(C=C3)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 198°C to 202°C
Quantity 250 mg
Formula Weight 397.45
Percent Purity 96%
Chemical Name or Material 4-[4-(2-Methylphenoxy)phenylsulfonylaminomethyl]benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'