missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ 300mL Tamper Evident Specimen Containers
These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples.
Brand: Simport™ C581-300
Additional Details : Weight : 2.50000kg
Description
- Container and closure are made of polypropylene
- Tight-fitting lids prevent leaks and odors
- Tamperproof lid is opened by first removing tab on one side of lid, then by lifting the cover
- Resists temperatures from −20° to +80°C
Specifications
| White | |
| Polypropylene | |
| White | |
| 45 mm | |
| 111 mm | |
| Yes | |
| Specimen container |
| Polypropylene | |
| 300 mL | |
| Hazardous specimens | |
| Lid | |
| 100/Cs | |
| Round |