missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiophenemethanol, 97%, ACROS Organics™
Brand: Acros Organics 442240050
Additional Details : Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
71637-34-8 | |
114.17 | |
3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol | |
thiophen-3-ylmethanol |
C5H6OS | |
BOWIFWCBNWWZOG-UHFFFAOYSA-N | |
123570 | |
C1=CSC=C1CO |
Specifications
3-Thiophenemethanol | |
100.5°C | |
73.0°C (6.0 Torr) | |
71637-34-8 | |
C5H6OS | |
BOWIFWCBNWWZOG-UHFFFAOYSA-N | |
thiophen-3-ylmethanol | |
123570 | |
97% |
1.2110g/mL | |
1.211 | |
5g | |
96% min. (GC) | |
3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol | |
C1=CSC=C1CO | |
114.17 | |
114.17 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Thiophenemethanol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.