missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrrolidin-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3485359
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
884507-42-0
C12H15NO
189.26
MFCD07782420
XHHOLQHPKIDQEM-UHFFFAOYSA-N
3-pyrrolidin-1-ylmethyl benzaldehyde, 3-pyrrolidin-1-yl methyl benzaldehyde, benzaldehyde, 3-1-pyrrolidinylmethyl, 1-3-formylbenzyl pyrrolidine, 3-pyrrolidinylmethyl benzaldehyde, 3-pyrrolidin-1-ylmethyl-benzaldehyde
18525870
3-(pyrrolidin-1-ylmethyl)benzaldehyde
O=CC1=CC(CN2CCCC2)=CC=C1
This item is not returnable. View return policy
884507-42-0
C12H15NO
189.26
MFCD07782420
XHHOLQHPKIDQEM-UHFFFAOYSA-N
3-pyrrolidin-1-ylmethyl benzaldehyde, 3-pyrrolidin-1-yl methyl benzaldehyde, benzaldehyde, 3-1-pyrrolidinylmethyl, 1-3-formylbenzyl pyrrolidine, 3-pyrrolidinylmethyl benzaldehyde, 3-pyrrolidin-1-ylmethyl-benzaldehyde
18525870
3-(pyrrolidin-1-ylmethyl)benzaldehyde
O=CC1=CC(CN2CCCC2)=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.26
Percent Purity 97%
Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'