missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinemethanol, 98%, Thermo Scientific Chemicals

Catalog Number p-4259057
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4606-65-9
C6H13NO
115.17
VUNPWIPIOOMCPT-UHFFFAOYSA-N
3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol
107308
piperidin-3-ylmethanol
C1CC(CNC1)CO
This item is not returnable. View return policy
4606-65-9
C6H13NO
115.17
VUNPWIPIOOMCPT-UHFFFAOYSA-N
3-piperidinemethanol, 3-hydroxymethyl piperidine, piperidin-3-yl-methanol, 3-hydroxymethylpiperidine, 3-piperidinylmethanol, piperidine-3-methanol, 3-piperidine methanol, 3-piperidylmethanol, 3-piperidinyl methanol, piperidin-3-yl methanol
107308
piperidin-3-ylmethanol
C1CC(CNC1)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Density 1.0200g/mL
Boiling Point 236.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21,II,8
Specific Gravity 1.02
Solubility Information Solubility in water: soluble
Formula Weight 115.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Piperidinemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Piperidinemethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'