Learn More
3-Phenyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™
Catalog Number p-4510014
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
1572-10-7
C9H9N3
159.192
PWSZRRFDVPMZGM-UHFFFAOYSA-N
3-phenyl-1h-pyrazol-5-amine, 3-amino-5-phenylpyrazole, 5-amino-3-phenylpyrazole, 1h-pyrazol-3-amine, 5-phenyl, 5-phenyl-2h-pyrazol-3-amine, pyrazole, 5-amino-3-phenyl, 3-amino-5-phenyl-1h-pyrazole, 5-phenyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-phenyl, 5-phenyl-1h-pyrazol-3-ylamine
136655
5-phenyl-1H-pyrazol-3-amine
C1=CC=C(C=C1)C2=CC(=NN2)N
1572-10-7
C9H9N3
159.192
PWSZRRFDVPMZGM-UHFFFAOYSA-N
3-phenyl-1h-pyrazol-5-amine, 3-amino-5-phenylpyrazole, 5-amino-3-phenylpyrazole, 1h-pyrazol-3-amine, 5-phenyl, 5-phenyl-2h-pyrazol-3-amine, pyrazole, 5-amino-3-phenyl, 3-amino-5-phenyl-1h-pyrazole, 5-phenyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-phenyl, 5-phenyl-1h-pyrazol-3-ylamine
136655
5-phenyl-1H-pyrazol-3-amine
C1=CC=C(C=C1)C2=CC(=NN2)N
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1H-pyrazol-3-amine