missing translation for 'onlineSavingsMsg'
Learn More

3-Morpholinobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-3110461
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
215309-00-5
C11H13NO3
207.229
MFCD06659078
VSKFQESEPGOWBS-UHFFFAOYSA-N
3-morpholinobenzoic acid, 3-4-morpholinyl benzoic acid, 3-morpholin-4-yl-benzoic acid, 3-morpholin-4-yl benzoic acid, 3-morpholinobenzoicacid, benzoic acid, 3-4-morpholinyl, 4-3-carboxyphenyl morpholine, pubchem19534, 3-4-morpholino benzoic acid
2795549
3-morpholin-4-ylbenzoic acid
C1COCCN1C2=CC=CC(=C2)C(=O)O
This item is not returnable. View return policy
215309-00-5
C11H13NO3
207.229
MFCD06659078
VSKFQESEPGOWBS-UHFFFAOYSA-N
3-morpholinobenzoic acid, 3-4-morpholinyl benzoic acid, 3-morpholin-4-yl-benzoic acid, 3-morpholin-4-yl benzoic acid, 3-morpholinobenzoicacid, benzoic acid, 3-4-morpholinyl, 4-3-carboxyphenyl morpholine, pubchem19534, 3-4-morpholino benzoic acid
2795549
3-morpholin-4-ylbenzoic acid
C1COCCN1C2=CC=CC(=C2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 3-Morpholinobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'