missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-146591
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
619-14-7
C7H5NO5
183.12
MFCD00007110
XLDLRRGZWIEEHT-UHFFFAOYSA-N
3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
69265
3-hydroxy-4-nitrobenzoic acid
OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
This item is not returnable. View return policy
619-14-7
C7H5NO5
183.12
MFCD00007110
XLDLRRGZWIEEHT-UHFFFAOYSA-N
3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
69265
3-hydroxy-4-nitrobenzoic acid
OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 233°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HOC6H3(NO2)CO2H
Quantity 25 g
Beilstein 10, 146
Solubility Information Solubility in water: insoluble
Formula Weight 183.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Hydroxy-4-nitrobenzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-4-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'