missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080789
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
537-73-5
C10H10O4
194.186
MFCD00004391
QURCVMIEKCOAJU-HWKANZROSA-N
isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid
736186
CHEBI:27794
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
COC1=C(C=C(C=C1)C=CC(=O)O)O
This item is not returnable. View return policy
537-73-5
C10H10O4
194.186
MFCD00004391
QURCVMIEKCOAJU-HWKANZROSA-N
isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid
736186
CHEBI:27794
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
COC1=C(C=C(C=C1)C=CC(=O)O)O

3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY
Melting Point ∼231°C (decomposition)
Quantity 1 g
Beilstein 2212760
Solubility Information Soluble in ethanol and methanol.
Formula Weight 194.19
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic acid, predominantly trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'