missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals

Catalog Number p-7027931
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
404-72-8
C7H4FNS
153.17
MFCD00004804
HDBACITVPQEAGG-UHFFFAOYSA-N
3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
9834
1-fluoro-3-isothiocyanatobenzene
FC1=CC=CC(=C1)N=C=S
This item is not returnable. View return policy
404-72-8
C7H4FNS
153.17
MFCD00004804
HDBACITVPQEAGG-UHFFFAOYSA-N
3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
9834
1-fluoro-3-isothiocyanatobenzene
FC1=CC=CC(=C1)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.27
Boiling Point 227°C
Flash Point 85°C (185°F)
Odor Pungent
Refractive Index 1.62
Quantity 5 g
UN Number UN2922
Beilstein 2081172
Sensitivity Moisture sensitive
Formula Weight 153.18
Percent Purity ≥97%
Chemical Name or Material 3-Fluorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'