missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-isocyanatothiophene, 95%, Thermo Scientific™

Catalog Number p-3586715
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
937795-88-5
C5H2BrNOS
204.041
MFCD09879904
HVJDEHSHJHNENX-UHFFFAOYSA-N
4-bromothiophen-3-isocyanate, 4-bromothien-3-yl isocyanate
24229539
3-bromo-4-isocyanatothiophene
C1=C(C(=CS1)Br)N=C=O
This item is not returnable. View return policy
937795-88-5
C5H2BrNOS
204.041
MFCD09879904
HVJDEHSHJHNENX-UHFFFAOYSA-N
4-bromothiophen-3-isocyanate, 4-bromothien-3-yl isocyanate
24229539
3-bromo-4-isocyanatothiophene
C1=C(C(=CS1)Br)N=C=O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.05
Percent Purity 95%
Chemical Name or Material 3-Bromo-4-isocyanatothiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'