missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals

Catalog Number p-58090
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
521-31-3
C8H7N3O2
177.16
MFCD00006890
HWYHZTIRURJOHG-UHFFFAOYSA-N
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
10638
5-amino-2,3-dihydrophthalazine-1,4-dione
C1=CC2=C(C(=C1)N)C(=O)NNC2=O
This item is not returnable. View return policy
521-31-3
C8H7N3O2
177.16
MFCD00006890
HWYHZTIRURJOHG-UHFFFAOYSA-N
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
10638
5-amino-2,3-dihydrophthalazine-1,4-dione
C1=CC2=C(C(=C1)N)C(=O)NNC2=O

3-Aminophthalhydrazide, 98%, pure, Acros Organics, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 319.0°C to 320.0°C
Color Blue-Purple to Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 25,IV,4192
Merck Index 15,566
Solubility Information (2% in 1 N NaOH) Clear yellow to yellow-green
Formula Weight 177.16
Percent Purity 98%
Grade Pure
Physical Form Powder
Chemical Name or Material 3-Aminophthalhydrazide, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'