missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrazole-1-acetic acid, 95%, Thermo Scientific™

Catalog Number p-7077651
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
16034-49-4
C7H9N2O2
153.16
MFCD00297249
JYSWEDYPQJOEPO-UHFFFAOYSA-M
2-3,5-dimethyl-1h-pyrazol-1-yl acetic acid, 3,5-dimethyl-pyrazol-1-yl-acetic acid, 3,5-dimethyl-1h-pyrazol-1-yl acetic acid, 3,5-dimethylpyrazol-1-yl acetic acid, 1h-pyrazole-1-acetic acid, 3,5-dimethyl, 3-nitro-benzene-thiol, 3,5-dimethyl-1h-pyrazole-1-acetic acid, 1h-pyrazole-1-aceticacid, 3,5-dimethyl, 2-3,5-dimethylpyrazolyl acetic acid, 3,5-dimethyl-pyrazol-1-yl acetic acid
572875
2-(3,5-dimethylpyrazol-1-yl)acetic acid
CC1=NN(CC([O-])=O)C(C)=C1
This item is not returnable. View return policy
16034-49-4
C7H9N2O2
153.16
MFCD00297249
JYSWEDYPQJOEPO-UHFFFAOYSA-M
2-3,5-dimethyl-1h-pyrazol-1-yl acetic acid, 3,5-dimethyl-pyrazol-1-yl-acetic acid, 3,5-dimethyl-1h-pyrazol-1-yl acetic acid, 3,5-dimethylpyrazol-1-yl acetic acid, 1h-pyrazole-1-acetic acid, 3,5-dimethyl, 3-nitro-benzene-thiol, 3,5-dimethyl-1h-pyrazole-1-acetic acid, 1h-pyrazole-1-aceticacid, 3,5-dimethyl, 2-3,5-dimethylpyrazolyl acetic acid, 3,5-dimethyl-pyrazol-1-yl acetic acid
572875
2-(3,5-dimethylpyrazol-1-yl)acetic acid
CC1=NN(CC([O-])=O)C(C)=C1
Melting Point 186.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 154.17
Percent Purity 95%
Chemical Name or Material 3,5-Dimethyl-1H-pyrazole-1-acetic acid
compliance-icons
Product Identifier
  • 3,5-Dimethyl-1H-pyrazole-1-acetic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'