Learn More
3,5-Difluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog Number p-4228600
Click to view available options
Quantity:
1 g
5 g
210532-25-5
C6H3ClF2O2S
212.59
MFCD02091378
IIQKIICAOXAXEJ-UHFFFAOYSA-N
3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
2774058
3,5-difluorobenzenesulfonyl chloride
FC1=CC(=CC(F)=C1)S(Cl)(=O)=O
210532-25-5
C6H3ClF2O2S
212.59
MFCD02091378
IIQKIICAOXAXEJ-UHFFFAOYSA-N
3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
2774058
3,5-difluorobenzenesulfonyl chloride
FC1=CC(=CC(F)=C1)S(Cl)(=O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 55.9°C to 58.2°C
Color Orange to Tan
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 212.6
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Difluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Difluorobenzenesulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.