missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ 3.3 Borosilicate Glass Burettes
Brand: VITLAB™ 105699
Additional Details : Weight : 0.80000kg
Specifications
0.1 mL | |
PMP, PTFE | |
50 mL | |
800 mm | |
Burette |
Schellbach Stripes | |
±0.1 mL | |
DIN ISO 384 | |
Borosilicate Glass |