missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-(2-Methoxyphenoxy)piperidine
Click to view available options
Quantity:
250 mg
1 g
Specifications
Quantity | 250 mg |
Formula Weight | 207.27 |
Chemical Name or Material | 3-(2-Methoxyphenoxy)piperidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'