missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-Pyrrol-1-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 202.28 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1H-pyrrol-1-yl)benzene-1-carbothioamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'