Learn More
Afora™ 3/14 Round Bottom Flask
Use Afora™ 3/14 Round Bottom Flask for chemical or biochemical work. Its spherical bottom makes it versatile piece of laboratory equipment.
Brand: Afora™ 3/14E
Additional Details : Weight : 0.04600kg
Description
- Weight: 0.046kg
- Precision measurement
- Glass
Specifications
Weight: 0.046kg | |
88 mm | |
Round |
Glass | |
1 Each | |
50 mm |