missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-1-[(1-Methylethyl)amino]-3-phenoxypropan-2-ol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(2RS)-1-[(1-Methylethyl)amino]-3-phenoxypropan-2-ol, Mikromol
Specifications
Specifications
| Chemical Name or Material | (2RS)-1-[(1-Methylethyl)amino]-3-phenoxy-propan-2-ol |
| CAS | 7695-63-8 |
| Analyte or Component Names | (2RS)-1-[(1-Methylethyl)amino]-3-phenoxy-propan-2-ol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H19 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Metoprolol Succinate Imp. F (EP), Metoprolol Tartrate Imp. F (EP), Metoprolol Imp. F (EP), (2RS)-1-[(1-Methylethyl)amino]-3-phenoxypropan-2-ol, Metoprolol Succinate Impurity F, Metoprolol Tartrate Impurity F, Metoprolol Impurity F, Metoprolol Impurity F |
| InChI Formula | InChI=1S/C12H19NO2/c1-10(2)13-8-11(14)9-15-12-6-4-3-5-7-12/h3-7,10-11,13-14H,8-9H2,1-2H3 |
| Solution Type | Neat |
| SMILES | CC(C)NCC(O)COc1ccccc1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction