missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan Isopropyl Ester Hydrochloride, Mikromol™

Product Code. 30049212
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30049212

Brand: Mikromol MM0172.190025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Tryptophan Isopropyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Tryptophan Isopropyl Ester Hydrochloride
CAS 71959-71-2
Analyte or Component Names L-Tryptophan Isopropyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H18 N2 O2 . Cl H
Certifications/Compliance ISO 9001
Synonym L-Tryptophan Isopropyl Ester Hydrochloride, L-Tryptophan, 1-methylethyl ester, hydrochloride (1:1), L-Tryptophan, 1-methylethyl ester, monohydrochloride (9CI)
InChI Formula InChI=1S/C14H18N2O2.ClH/c1-9(2)18-14(17)12(15)7-10-8-16-13-6-4-3-5-11(10)13;/h3-6,8-9,12,16H,7,15H2,1-2H3;1H/t12-;/m0./s1
SMILES Cl.CC(C)OC(=O)[C@@H](N)Cc1c[nH]c2ccccc12
IUPAC Name propan-2-yl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Molecular Weight (g/mol) 282.77
Quantity 25 mg
Formula Weight 282.1135 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.