Learn More
2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride, 97%, ACROS Organics™
Brand: Acros Organics 428240010
Additional Details : CAS : 82495-75-8 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
82495-75-8 | |
429.01 | |
2-trimethylsilyl ethoxymethyltriphenylphosphonium chloride, sem-triphenylphosphonium chloride, 2-trimethylsilyl ethoxymethyl triphenylphosphonium chloride, phosphonium, triphenyl 2-trimethylsilyl ethoxy methyl-, chloride, acmc-209pp9, 2-trimethylsilyl ethoxymethyltriphenylphosphoniumchloride, 2-trimethylsilyl ethoxymethyl-triphenylphosphonium chloride, 2-trimethylsilyl-ethoxymethyl triphenylphosphonium chloride, triphenyl-2-trimethylsilanyl-ethoxymethyl phosphonium chloride | |
triphenyl(2-trimethylsilylethoxymethyl)phosphanium;chloride |
C24H30ClOPSi | |
NEUMNYXEDIPGJD-UHFFFAOYSA-M | |
11396265 | |
C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-] |
Specifications
2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride | |
166.0°C | |
82495-75-8 | |
C24H30ClOPSi | |
2-trimethylsilyl ethoxymethyltriphenylphosphonium chloride, sem-triphenylphosphonium chloride, 2-trimethylsilyl ethoxymethyl triphenylphosphonium chloride, phosphonium, triphenyl 2-trimethylsilyl ethoxy methyl-, chloride, acmc-209pp9, 2-trimethylsilyl ethoxymethyltriphenylphosphoniumchloride, 2-trimethylsilyl ethoxymethyl-triphenylphosphonium chloride, 2-trimethylsilyl-ethoxymethyl triphenylphosphonium chloride, triphenyl-2-trimethylsilanyl-ethoxymethyl phosphonium chloride | |
C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-] | |
429.01 | |
429.01 |
Glass bottle | |
1g | |
97% | |
(CH3)3Si(CH2)3OCH2P(C6H5)3Cl | |
NEUMNYXEDIPGJD-UHFFFAOYSA-M | |
triphenyl(2-trimethylsilylethoxymethyl)phosphanium;chloride | |
11396265 | |
97% |
Safety and Handling
- 2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.