Learn More
2-(Tetrazol-5-yl)phenylboronic acid, 95%, Thermo Scientific Chemicals
Catalog Number 11906761
Click to view available options
Quantity:
1 g
5 g
155884-01-8
C7H7BN4O2
189.97
MFCD06796325
GVRXWYFECKHTSJ-UHFFFAOYSA-N
2-5-tetrazolyl phenylboronic acid, 2-1h-tetrazol-5-yl phenyl boronic acid, 2-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl-phenylboronic acid, 2-1h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenyl boronic acid, boronic acid, 2-1h-tetrazol-5-yl phenyl, boronic acid,b-2-2h-tetrazol-5-yl phenyl
11171482
[2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
OB(O)C1=CC=CC=C1C1=NNN=N1
155884-01-8
C7H7BN4O2
189.97
MFCD06796325
GVRXWYFECKHTSJ-UHFFFAOYSA-N
2-5-tetrazolyl phenylboronic acid, 2-1h-tetrazol-5-yl phenyl boronic acid, 2-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl-phenylboronic acid, 2-1h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenyl boronic acid, boronic acid, 2-1h-tetrazol-5-yl phenyl, boronic acid,b-2-2h-tetrazol-5-yl phenyl
11171482
[2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
OB(O)C1=CC=CC=C1C1=NNN=N1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 148°C to 152°C
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 189.97
Percent Purity 95%
Chemical Name or Material 2-(Tetrazol-5-yl)phenylboronic acid
compliance-icons
Product Identifier
  • 2-(Tetrazol-5-yl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.